missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYPL2 Polyclonal Antibody

Description
A suggested positive control is human spleen tissue lysate. PA5-20776 can be used with blocking peptide PEP-0890.
SYPL2, also known as Mitsugumin 29, was initially identified as a transmembrane protein from the triad junction in skeletal muscle that had significant homology with members of the synaptophysin family. SYPL2 is thought to participate in the excitation-contraction coupling process of skeletal muscle as SYPL2-null mice showed reduced muscle contractile force and altered triad junction structure and increased susceptibility to fatigue of the skeletal muscle. SYPL2 plays a critical role in muscle Ca2+ signaling by regulating the process of store-operated Ca2+ entry and interacts with ryanodine receptor (RyR), thereby influencing intracellular Ca2+ homeostasis through changes in the RyR/Ca2+ release function. Co-expression of SYPL2 and RyR in cultured cells leads to apoptotic cell death resulting from the depletion of Ca2+ from the intracellular stores.
Specifications
Specifications
| Antigen | SYPL2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SYPL2 |
| Gene Accession No. | O89104, Q5VXT5 |
| Gene Alias | AI552439; MG29; mitsugumin 29; mitsugumin29; mitsugumin-29; synaptophysin like 2; synaptophysin-like 2; synaptophysin-like protein 2; Sypl2; UNQ6348 |
| Gene Symbols | SYPL2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?