missing translation for 'onlineSavingsMsg'
Learn More

SYPL1 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18402338 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402338 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402338 Supplier Novus Biologicals Supplier No. NBP176316G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SYPL1 Polyclonal antibody specifically detects SYPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYPL1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias H-SP1, Pantophysin, synaptophysin-like 1, synaptophysin-like protein, synaptophysin-like protein 1, SYPL
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SYPL1. The immunogen is located within the first 50 amino acids of SYPL1. Amino Acid Squence: APNIYLVRQRISRLGQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6856
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.