missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYPL1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76316B
This item is not returnable.
View return policy
Description
SYPL1 Polyclonal antibody specifically detects SYPL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| SYPL1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Biotin | |
| H-SP1, Pantophysin, synaptophysin-like 1, synaptophysin-like protein, synaptophysin-like protein 1, SYPL | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SYPL1. The immunogen is located within the first 50 amino acids of SYPL1. Amino Acid Squence: APNIYLVRQRISRLGQ | |
| 0.1 mL | |
| Neuroscience | |
| 6856 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction