missing translation for 'onlineSavingsMsg'
Learn More

SYP-5, MedChemExpress

Product Code. 16499676
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499676 50 mg 50mg
16474416 100 mg 100mg
16465857 5 mg 5mg
16433128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499676 Supplier Medchem Express Supplier No. HY100693

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SYP-5 is a novel HIF-1 inhibitor, suppresses tumor cells invasion and angiogenesis.

SYP-5 is a novel HIF-1 inhibitor, suppresses tumor cells invasion and angiogenesis.

Specifications

Chemical Name or Material SYP-5
CAS 1384268-04-5
Color Orange
Purity Grade Notes Research
Molecular Formula C18H16O3S
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (106.70 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C(C=CC(C)(C)O2)=C2C=C1)/C=C/C3=CC=CS3
Molecular Weight (g/mol) 312.38
Formula Weight 312.38
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.