missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syntrophin alpha-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114304
This item is not returnable.
View return policy
Description
Syntrophin alpha-1 Polyclonal Antibody for Western Blot
Syntrophin alpha-1 (Dystrophin, SNTA1) is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.
Specifications
| Syntrophin alpha-1 | |
| Polyclonal | |
| Unconjugated | |
| SNTA1 | |
| 59 kDa dystrophin-associated protein A1 acidic component 1; 59-1 DAP; acidic alpha 1 syntrophin; alpha-1-syntrophin; alpha1-syntrophin; alpha-syntrophin; AW228934; dJ1187J4.5; dystrophin-associated protein A1, 59kDa, acidic component; LQT12; pro-TGF-alpha cytoplasmic domain-interacting protein 1; RP23-192L3.1; SNT1; SNTA1; syntrophin alpha 1; syntrophin, acidic 1; syntrophin, alpha 1; syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component); syntrophin-1; TACIP1; Unknown (protein for MGC:142519) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20648, 362242, 6640 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q13424, Q61234 | |
| SNTA1 | |
| Synthetic peptide within mouse Syntrophin alpha 1 aa 174-223. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction