missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntrophin alpha 1 Antibody (OTI1H10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74422AF647
This item is not returnable.
View return policy
Description
Syntrophin alpha 1 Monoclonal specifically detects Syntrophin alpha 1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Syntrophin alpha 1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SNTA1 | |
| Full length recombinant protein of human Syntrophin alpha 1 (NP_003089) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1H10 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin | |
| Mouse | |
| Affinity Purified | |
| Cytoskeleton Markers, Neuroscience, Vision | |
| 6640.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction