missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntrophin alpha 1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1346
This item is not returnable.
View return policy
Description
Syntrophin alpha 1 Polyclonal specifically detects Syntrophin alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Syntrophin alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| 59 kDa dystrophin-associated protein A1 acidic component 1, dJ1187J4.5, LQT12, Pro-TGF-alpha cytoplasmic domain-interacting protein 1, SNT1alpha-1-syntrophin, syntrophin, alpha 1 (dystrophin-associated protein A1, 59kD, acidic component), syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component), syntrophin-1, TACIP1acidic alpha 1 syntrophin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_003089. | |
| SNTA1 | |
| Peptide with sequence ASGRRAPRTGLLE-C corresponding to N-Terminus according to NP_003089.1. | |
| 0.1 mg | |
| Cytoskeleton Markers, Neuroscience, Vision | |
| 6640 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction