missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Synthetic Human Ubiquitin Vinyl Sulfone Protein, CF
Description
Recombinant Human Ubiquitin-Vinyl Sulfone added to in vitro assays will inhibit deubiquitinating enzymes. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human Ubiquitin-Vinyl Sulfone concentration of 1-5 μM.
Specifications
Specifications
| Accession Number | P0CG47.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a solution in MES and DMSO. |
| Gene ID (Entrez) | 6233 |
| Quantity | 50 μg |
| Source | Chemically Synthesized Ubiquitin protein Met1 - Gly75 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Gene Alias | RPS27A, UBA52, UBB, UBB ubiquitin B, UBC |
| Conjugate | Vinyl Sulfone |
| Purity or Quality Grade | >95%, by HPLC. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?