missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Synthetic Disinfectant Wipes Zero Waste Box™
GREENER_CHOICE

Product Code. p-7249627
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy

Product Code. 17805935

Brand: Terracycle™ CS175SUK

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Recycle Synthetic Disinfectant Wipes using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

What you can recycle:

  • Synthetic Disinfectant Wipes

This box does not accept:

  • Wipe Packaging
  • Other Cleaning Supplies
  • Natural Disinfectant Wipes

Specifications

Capacity (Metric) 7 kg
Capacity (English) 15 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 25 x 25 x 46 cm
Disposable Single-use
Latex Free Yes
Size Small
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.