Learn More
PURUS™ Synthetic Cleanroom Paper
Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.
Brand:  PURUS™ LCIW 2046C
Additional Details : Weight : 12.50000kg
 View more versions of this product
Product Code. 16430051
  • £215.00 / Pack of 5000
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Propriatary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Produced in ISO 4 (Class 10) cleanroom
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Specifications
Synthetic
White
Autoclavable
Yes
250 Sheets/Pk., 20 Packs/Cs.
80 gsm
No
210 x 148 mm
For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Cleanroom Paper
A5