missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syntenin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121564
This item is not returnable.
View return policy
Description
Syntenin 2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
SDCBP2 contains 2 PDZ (DHR) domains. The function of SDCBP2 remains unknown.
Specifications
| Syntenin 2 | |
| Polyclonal | |
| Unconjugated | |
| SDCBP2 | |
| SDCBP2; Similar to TACIP18; SITAC; SITAC18; ST-2; syndecan binding protein (syntenin) 2; syndecan binding protein 2; syndecan-binding protein 2; Syntenin-2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 228765, 27111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q99JZ0, Q9H190 | |
| SDCBP2 | |
| Recombinant protein Syntenin 2. The antigen corresponds to amino acid range 1-292 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction