missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syntenin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582846
This item is not returnable.
View return policy
Description
Immunogen sequence: ANVAVVSGAP LQGQLVARPS SINYMVAPVT GNDVGIRRAE IKQGIREVIL CKDQDGKIGL RLKSI Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 89%.
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| Syntenin 1 | |
| Polyclonal | |
| Unconjugated | |
| SDCBP | |
| MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6386 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00560 | |
| SDCBP | |
| Recombinant protein corresponding to Human SDCBP. Recombinant protein control fragment (Product #RP-93800). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction