missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntenin-1 Mouse anti-Human, Clone: 2B1A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68096-1-IG-20UL
This item is not returnable.
View return policy
Description
Syntenin-1 Monoclonal antibody specifically detects Syntenin-1 in Human samples. It is validated for IHC, WB, ELISASpecifications
| Syntenin-1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Mouse | |
| 20 μL | |
| 6386 | |
| Store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, ELISA | |
| 2B1A9 | |
| Unconjugated | |
| BC092414 | |
| Protein G purification | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction