missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntenin 1 Antibody (K10P3D5*H3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50287-0.025ml
This item is not returnable.
View return policy
Description
Syntenin 1 Monoclonal antibody specifically detects Syntenin 1 in Human samples. It is validated for Western Blot
Specifications
| Syntenin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000 | |
| MDA9, MDA-9, melanoma differentiation associated protein-9, Melanoma differentiation-associated protein 9, Pro-TGF-alpha cytoplasmic domain-interacting protein 18, Scaffold protein Pbp1, ST1, SYCLTACIP18, syndecan binding protein (syntenin), Syndecan-binding protein 1, syntenin-1 | |
| Peptide - HIIKRMAPS | |
| 0.025 mL | |
| Cell Biology, Cytokines and Growth Factors, Immunology, Neuroscience | |
| 6386 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| K10P3D5*H3 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction