missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin-BP1 Rabbit anti-Human, Mouse, Rat, Clone: S05-4B6, Novus Biologicals™
Rabbit Monoclonal Antibody
£232.00 - £451.00
Specifications
| Antigen | Syntaxin-BP1 |
|---|---|
| Clone | S05-4B6 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18320955
|
Novus Biologicals
NBP3-15097-25UL |
25 μg |
£232.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18371855
|
Novus Biologicals
NBP3-15097-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntaxin-BP1 Monoclonal antibody specifically detects Syntaxin-BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Syntaxin-BP1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EIEE4, FLJ37475, hUNC18, MUNC18-1, N-Sec1, NSEC1, P67, Protein unc-18 homolog 1, Protein unc-18 homolog A, RBSEC1, syntaxin binding protein 1, syntaxin-binding protein 1, Unc18-1, unc-18A, UNC18A, UNC18neuronal SEC1 | |
| A synthetic peptide of human Syntaxin-BP1 (Uniprot#: P61764) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S05-4B6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 6812 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title