Learn More
Syntaxin 7 Antibody (CL0257) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52893H
Product Code. 30305245
  • £558.00 / 0.10mL
Description
Syntaxin 7 Monoclonal antibody specifically detects Syntaxin 7 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Syntaxin 7
Monoclonal
HRP
syntaxin 7, syntaxin-7
This antibody was developed using a recombinant protein derived from O15400, with the exact immunogen sequence remaining proprietary.
0.1 mL
Endosome Markers
8417
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0257
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified