missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syntaxin 4 Recombinant Rabbit Monoclonal Antibody (K01_1Y96)
Description
Syntaxin 4 Recombinant Monoclonal Antibody for Western Blot
STX4 is a plasma membrane t-SNARE that mediates docking of transport vesicles.STX4 is necessary for the translocation of SLC2A4 from intracellular vesicles to the plasma membrane. Together with STXB3 and VAMP2, STX4 may also play a role in docking/fusion of intracellular GLUT4-containing vesicles with the cell surface in adipocytes. STX4 may also play a role in docking of synaptic vesicles at presynaptic active zones.
Specifications
Specifications
| Antigen | Syntaxin 4 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1Y96 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | STX4 |
| Gene Accession No. | P70452, Q08850, Q12846 |
| Gene Alias | hypothetical protein LOC508675; p35-2; Renal carcinoma antigen NY-REN-31; Stx4; STX4A; Syn4; Syn-4; syntaxin 4; syntaxin 4A (placental); syntaxin-4; syntaxin-4 isoform A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?