missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 4 Polyclonal Antibody

Brand: Thermo Scientific PA5118290
This item is not returnable.
View return policy
Description
Specifications
| Syntaxin 4 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q12846 | |
| STX4 | |
| E. coli-derived Human STX4 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| STX4 | |
| hypothetical protein LOC508675; p35-2; Renal carcinoma antigen NY-REN-31; Stx4; STX4A; Syn4; Syn-4; syntaxin 4; syntaxin 4A (placental); syntaxin-4; syntaxin-4 isoform A | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 6810 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction