missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 3 Rabbit anti-Human, Mouse, Rat, Clone: 3L9Y9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Syntaxin 3 |
|---|---|
| Clone | 3L9Y9 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319681
|
Bio-Techne
NBP3-16260-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18399665
|
Novus Biologicals
NBP3-16260-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntaxin 3 Monoclonal antibody specifically detects Syntaxin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Syntaxin 3 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ30906, STX3Asyntaxin 3A, syntaxin 3, syntaxin-3 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Syntaxin 3 (Q13277). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3L9Y9 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6809 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title