missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syntaxin 2 Monoclonal Antibody (338930)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524068
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human Epimorphin is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
The product of this gene belongs to the syntaxin/epimorphin family of proteins. The syntaxins are a large protein family implicated in the targeting and fusion of intracellular transport vesicles. The product of this gene regulates epithelial-mesenchymal interactions and epithelial cell morphogenesis and activation. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Syntaxin 2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q00262 | |
| Stx2 | |
| E. coli-derived recombinant mouse Epimorphin/Syntaxin 2 Met1-Arg189. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen), Western Blot | |
| 338930 | |
| Unconjugated | |
| Stx2 | |
| AW538950; EPIM; Epimorphin; EPM; G1-536-1; I79_024396; MGC51014; repro34; Stx2; STX2A; STX2B; STX2C; Syn-2; syntaxin 2; syntaxin-2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 13852 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction