missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17815-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.Specifications
| Syntaxin 17 | |
| Polyclonal | |
| Unconjugated | |
| STX17 | |
| STX17, syntaxin 17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 252853, 55014, 67727 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56962, Q9D0I4, Q9Z158 | |
| STX17 | |
| Syntaxin 17 Fusion Protein Ag12134 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction