missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 16 Antibody (S09-9A8), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | Syntaxin 16 |
|---|---|
| Clone | S09-9A8 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655243
|
Novus Biologicals
NBP3-19783-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637603
|
Novus Biologicals
NBP3-19783-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntaxin 16 Monoclonal antibody specifically detects Syntaxin 16 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Syntaxin 16 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 8675 | |
| IgG | |
| Affinity purified |
| S09-9A8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| MGC90328, Syn16, SYN16hsyn16, syntaxin 16, syntaxin-16 | |
| A synthetic peptide of human Syntaxin 16 (Uniprot # O14662) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title