missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Syntaphilin Polyclonal antibody specifically detects Syntaphilin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Syntaphilin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | bA314N13.5, KIAA0374, MGC46096, syntaphilin |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 16 amino acid synthetic peptide from near the center of human Syntaphilin. The immunogen is located within amino acids 350 - 400 of Syntaphilin. Amino Acid Squence: VGDELEAPEPITRGPT |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?