missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaphilin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | Syntaphilin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Syntaphilin Polyclonal specifically detects Syntaphilin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Syntaphilin | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O15079 | |
| 9751 | |
| Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human Syntaphilin. The immunogen is located within amino acids 80 - 130 of Syntaphilin. Amino Acid Squence: KARLKDTQDRLQDRDTEI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| bA314N13.5, KIAA0374, MGC46096, syntaphilin | |
| SNPH | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title