missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntabulin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16972-1-AP-150UL
This item is not returnable.
View return policy
Description
Syntabulin/GOLSYN is part of a kinesin motor-adaptor complex that is critical for the anterograde axonal transport of active zone components and contributes to activity-dependent presynaptic assembly during neuronal development.Specifications
| Syntabulin | |
| Polyclonal | |
| Unconjugated | |
| SYBU | |
| Golgi localized protein, GOLSYN, KIAA1472, SYBU, Syntabulin, Syntaxin 1 binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319613, 500865, 55638 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BHS8, Q9NX95 | |
| Sybu | |
| Syntabulin Fusion Protein Ag9344 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction