missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Synsation Gloves 4001 PF
Shop All Ansell Products
Click to view available options
Size:
Large, 8.5/9
Medium, 7.5/8
Small, 6.5/7
X-Large, 9.5/10
X-Small, 5.5/6
Unit Size:
Pack of 100
Description
- Quick and easy donning
- Beaded cuff to prevent cuff roll down
- Protects from latex Type I allergy
Specifications
Specifications
| Certifications/Compliance (Europe) | CE 0493, Category III, EN 16523-1, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 9001, ISO 13485, ISO 10993-10:2002 |
| Chemotherapeutic Rated | No |
| Accelerator Free | Yes |
| Applications | Healthcare |
| AQL | 1.5 |
| Color | Clear |
| Disposable | Single-use |
| Exterior Finish | Smooth |
| Finger Thickness (English) | 3.9 mil |
| Finger Thickness (Metric) | 0.1 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction