missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Quick and easy donning
- Beaded cuff to prevent cuff roll down
- Protects from latex Type I allergy
Specifications
Specifications
| Certifications/Compliance (Europe) | CE 0493, Category III, EN 16523-1, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, ISO 9001, ISO 13485, ISO 10993-10:2002 |
| Chemotherapeutic Rated | No |
| Accelerator Free | Yes |
| Applications | Healthcare |
| AQL | 1.5 |
| Color | Clear |
| Disposable | Single-use |
| Exterior Finish | Smooth |
| Finger Thickness (English) | 3.9 mil |
| Finger Thickness (Metric) | 0.1 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?