missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNPO2L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21480-1-AP-150UL
This item is not returnable.
View return policy
Description
SYNPO2L was initially identified as a novel heart-enriched gene that encodes a cytoskeletal protein highly expressed in the Z-disc of heart and skeletal muscle, associates with actin and interacts with a-actinin. It is a member of the synaptopodin family, sharing greatest homology with Synaptopodin 2. Recent studies have shown that SYNPO2L, while primarily localized to the sarcomere, can also translocate to the nucleus. A knockdown of SYNPO2L in zebrafish resulted in aberrant cardiac and skeletal muscle development and function, suggesting that it is a critical component of the sarcomere and plays an important role in muscle development.Specifications
| SYNPO2L | |
| Polyclonal | |
| Unconjugated | |
| SYNPO2L | |
| synaptopodin 2 like, Synaptopodin 2 like protein, SYNPO2L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305675, 68760, 79933 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BWB1, Q9H987 | |
| SYNPO2L | |
| SYNPO2L Fusion Protein Ag15683 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction