missing translation for 'onlineSavingsMsg'
Learn More

SYNPO2L Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18416088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416088 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416088 Supplier Novus Biologicals Supplier No. NBP282021AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SYNPO2L Polyclonal antibody specifically detects SYNPO2L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNPO2L
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Synaptopodin 2-Like, Synaptopodin 2-Like Protein
Host Species Rabbit
Immunogen SYNPO2L antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human SYNPO2L. The immunogen is located within amino acids 140 - 190 of SYNPO2L. Amino Acid Squence: PATQEKPRRPRRRGPTRPT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79933
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.