missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYNM Monoclonal Antibody (OTI3D10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526801
This item is not returnable.
View return policy
Description
SYNM Monoclonal Antibody for Western Blot, IHC (P)
Type-VI intermediate filament (IF) which plays an important cytoskeletal role within the muscle cell cytoskeleton. It forms heteropolymeric IFs with desmin and/or vimentin, and via its interaction with cytoskeletal proteins alpha-dystrobrevin, dystrophin, talin-1, utrophin and vinculin, is able to link these heteropolymeric IFs to adherens-type junctions, such as to the costameres, neuromuscular junctions, and myotendinous junctions within striated muscle cells.
Specifications
| SYNM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15061 | |
| SYNM | |
| Human recombinant protein fragment corresponding to amino acids 329-580 of SYNM produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D10 | |
| Unconjugated | |
| SYNM | |
| 4930412K21Rik; AI852401; Desmuslin; DMN; E130104F11; KIAA0353; SYN; synemin; synemin alpha; synemin beta; synemin, intermediate filament protein; synm | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23336 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction