missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synip Monoclonal Antibody (J.696.2)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514940
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
STXBP4 plays a role in the translocation of transport vesicles from the cytoplasm to the plasma membrane. STXBP4 inhibits the translocation of SLC2A4 from intracellular vesicles to the plasma membrane by STX4A binding and preventing the interaction between STX4A and VAMP2. Stimulation with insulin disrupts the interaction with STX4A, leading to increased levels of SLC2A4 at the plasma membrane. STXBP4 may also play a role in the regulation of insulin release by pancreatic beta cells after stimulation by glucose.
Specifications
| Synip | |
| Monoclonal | |
| Unconjugated | |
| Stxbp4 | |
| 6030470M02Rik; FLJ16496; MGC149829; MGC50337; STX4-interacting protein; Stxbp4; Synip; syntaxin 4 interacting protein; syntaxin 4-interacting protein; syntaxin binding protein 4; syntaxin-binding protein 4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20913, 252983, 303443 | |
| -20°C | |
| Liquid |
| Western Blot | |
| J.696.2 | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q6ZWJ1, Q9WV89 | |
| Stxbp4 | |
| Synthetic peptide corresponding to the sequence of human Synip. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction