missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYNGR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534231
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human (100%) Gibbon, Monkey, Marmoset (82%).
Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear. SYNGR3 is predominantly expressed in brain and placenta. The exact function of SYNGR2 is unknown, but studies suggest that it may play a role in regulating membrane traffic in non-neuronal cells.
Specifications
| SYNGR2 | |
| Polyclonal | |
| Unconjugated | |
| SYNGR2 | |
| cellugyrin; Clast2; MGC102914; synaptogyrin 2; synaptogyrin-2; Syngr2; UNQ352/PRO615 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9144 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O43760 | |
| SYNGR2 | |
| Synthetic 17 amino acid peptide from N-Terminus of human SYNGR2 / Synaptogyrin 2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction