missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYNGR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520877
This item is not returnable.
View return policy
Description
A suggested positive control is human lung tissue lysate. PA5-20877 can be used with blocking peptide PEP-0991.
Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear. SYNGR3 is predominantly expressed in brain and placenta. The exact function of SYNGR2 is unknown, but studies suggest that it may play a role in regulating membrane traffic in non-neuronal cells.
Specifications
| SYNGR2 | |
| Polyclonal | |
| Unconjugated | |
| SYNGR2 | |
| cellugyrin; Clast2; MGC102914; synaptogyrin 2; synaptogyrin-2; Syngr2; UNQ352/PRO615 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89815, 9144 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O43760, O54980 | |
| SYNGR2 | |
| A 17 amino acid peptide from near the amino terminus of human SYNGR2. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction