Learn More
Syngene™ Visible Light Converter
Shop All Syngene Products
Click to view available options
For Use With (Equipment):
G:BOX Systems
IG/UG and DG Systems
Unit Size:
Each
Description
- Use the Visible light converter in conjunction with a UV transilluminator provides a white surface for visualising visible protein gels.
Specifications
Specifications
| Intensity | Fixed |
| Wavelength | 302 nm |
| Description | UV (White) |
| For Use With (Equipment) | IG/UG and DG Systems |
| Type | Converter Plate |