Learn More
Syngene™ UV Transilluminator 2020S
Syngene™ UV Transilluminator 2020SUV is for illumination of any sample excited by UV light.
Brand: Syngene™ BT-2020S-E
This item is not returnable.
View return policy
Description
2020S 230V 50Hz 230W 241mm x 315mm x 80mm 254nm wavelength 200mm x 200mm filter area for benchtop use SyngeneSpecifications
| UV Transilluminator | |
| 230 w | |
| 241 x 315 x 80 mm | |
| 200 x 200 mm | |
| 230 V | |
| Fixed |
| Benchtop use | |
| 200 x 200 mm | |
| 230 V 50 Hz 230 w | |
| Syngene 2020S | |
| 50 Hz | |
| 254 nm |