missing translation for 'onlineSavingsMsg'
Learn More

Synoptics™ Syngene™ Blue Light Converter Screen
GREENER_CHOICE

Product Code. 15799489 Shop All Syngene Products
Change view
Click to view available options
Size:
25 x 30cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15799489 25 x 30cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15799489 Supplier Synoptics™ Supplier No. CONVERTBLUE2530
Add to basket
Add to basket
This item is not returnable. View return policy

Convert UV to blue light for imaging of Safe Dyes - use with Uv transilluminators

  • UV-to-blue light converter screen
  • Area 25 x 30cm

convertor, uv to blue, blue screen

Specifications

For Use With (Application) Safe dyes
Product Type Convertor Screen
Quantity 1Each
Color Blue
For Use With (Equipment) Syngene Gbox
Size 25 x 30cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.