missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNGAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19739-1-AP-20UL
This item is not returnable.
View return policy
Description
Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.Specifications
| SYNGAP1 | |
| Polyclonal | |
| Unconjugated | |
| SYNGAP1 | |
| KIAA1938, MRD5, Neuronal RasGAP, RASA1, RASA5, Synaptic Ras GAP 1, SYNGAP, SYNGAP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 240057, 8831 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F6SEU4, Q96PV0 | |
| SYNGAP1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction