missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SynGAP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537910
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Synaptic Ras-GTPase activating protein (SynGAP) is a PDZ-domain interacting protein expressed almost exclusively in the brain. SynGAP has been shown to be a major component of postsynaptic density and associates with post synaptic density 95 kDa (PSD-95), a synaptic scaffold protein, and synaptic N-methyl-D-aspartate (NMDA) receptors. SynGAP Ras-GTPase activity has also been shown to be inhibited by phosphorylation by CaM kinase II, another abundant protein located in the postsynaptic density. It has been proposed that SynGAP may function as a negative regulator of MAP Kinase activation in the absence of active NMDA receptors.
Specifications
| SynGAP | |
| Polyclonal | |
| Unconjugated | |
| SYNGAP1 | |
| DKFZp761G1421; Gm1963; KIAA1938; MRD5; Neuronal R; Neuronal RasGAP; OTTHUMP00000209046; p135 SynGAP; Ras GTPase-activating protein SynGAP; ras/Rap GTPase-activating protein SynGAP; RASA1; RASA5; synaptic Ras GTPase activating protein 1; synaptic Ras GTPase activating protein 1 homolog; synaptic Ras GTPase activating protein 1 homolog (rat); synaptic Ras GTPase activating protein, 135kDa; synaptic Ras GTPase-activating protein 1; Synaptic Ras-GAP 1; SYNGAP; Syngap1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 240057, 8831 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| F6SEU4, Q96PV0 | |
| SYNGAP1 | |
| Peptide with sequence C-ESAHIEREEYKLKEY, from the internal region near the C-terminus of SYNGAP1 (aa1169-1183). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction