Learn More
Agilent BioTek Synergy HTX Qualification Package - IQ/OQ/PQ
Product Code. 16552132 Shop All Agilent BioTek Products
Click to view available options
:
Synergy HTX Qualification Package - IQ/OQ/PQ
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
Product Code. 16552132
Brand: Agilent BioTek 1340508
Note:
Product qualification packages include thorough procedures to allow a user to perform installation qualification, operational qualification and performance qualification. Periodic maintenance procedures are also included to assist in maintaining the instruments for optimal performance. Product qualification packages contain only documentation and procedures. Many of these procedures require the use of QC test solutions or other accessories, but these are not included with the product qualification packages.
For Synergy HTX microplate reader models
  • Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
  • Comes on USB flash drive with the following contents:
    • Detailed required material list
    • Installation qualification procedure and checklist
    • Operational qualification procedure
    • Performance qualification procedure
    • Microsoft Excel spreadsheets for calculations required by procedures
    • Maintenance procedures
    • 5 year logbook for documenting all procedures/results
      • All of the documentation is available in both .doc and .pdf file format
Note:
  • Absorbance test plate (7260522) may be required in order to run some procedures specified for absorbance and multi-mode readers, but is not included and must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Synergy HTX microplate reader models
Includes Detailed required material list, Installation qualification procedure and checklist, Operational qualification procedure, Performance qualification procedure, Microsoft Excel spreadsheets for calculations, Maintenance procedures, 5 year logbook