missing translation for 'onlineSavingsMsg'
Learn More

SYNE1 Rabbit anti-Human, Mouse, Rat, Clone: 1D4D9, Novus Biologicals™

Product Code. 18377982 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377982 100 μg 100µL
18361822 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18377982 Supplier Novus Biologicals Supplier No. NBP315911100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SYNE1 Monoclonal antibody specifically detects SYNE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SYNE1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1D4D9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ARCA1, C6orf98, CPG2, dJ45H2.2, EC 4.2.1.49, EC 4.3.1.3, EDMD4, Enaptin, KIAA0796chromosome 6 open reading frame 98, KIAA1262FLJ41140, KIAA17568B, Myne-1, MYNE1DKFZp781J13156, Myocyte nuclear envelope protein 1, nesprin 1, nesprin-1, Nuclear envelope spectrin repeat protein 1, SCAR8FLJ30878, spectrin repeat containing, nuclear envelope 1, Synaptic nuclear envelope protein 1, synaptic nuclei expressed gene 1, Syne-1, SYNE-1B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 8200-8400 of human SYNE1 (Q8NF91).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 23345
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.