missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndetin/CCDC132 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27749-1-AP-20UL
This item is not returnable.
View return policy
Description
CCDC132 is a protein coding gene.Specifications
| Syndetin/CCDC132 | |
| Polyclonal | |
| Unconjugated | |
| VPS50 | |
| CCDC132, Syndetin, VPS50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 312083, 55610, 73288 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| F1LSG8, Q8CI71, Q96JG6 | |
| Vps50 | |
| Syndetin/CCDC132 Fusion Protein Ag26944 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction