missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Syndecan 3 Recombinant Rabbit Monoclonal Antibody (24GB3890)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557275
This item is not returnable.
View return policy
Description
Syndecan 3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene belongs to the syndecan proteoglycan family. It may play a role in the organization of cell shape by affecting the actin cytoskeleton, possibly by transferring signals from the cell surface in a sugar-dependent mechanism. Allelic variants of this gene have been associated with obesity.
Specifications
| Syndecan 3 | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75056, P33671, Q64519 | |
| Sdc3 | |
| A synthesized peptide derived from human Syndecan 3 (aa 51-78). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3890 | |
| Unconjugated | |
| Sdc3 | |
| Kiaa0468; mKIAA0468; Neuroglycan; N-syndecan; SDC3; SDCN; syn-3; Synd3; syndecan 1; syndecan 3; syndecan neural type; syndecan proteoglycan 3; syndecan-3; wu:fa97a01 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116673, 20970, 9672 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction