missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Syndecan 2 Polyclonal Antibody

Product Code. 18045721
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18045721 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18045721 Supplier Invitrogen™ Supplier No. PA5144961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KVETTTLNIQ NKIPAQTKSP EETDKEKVHL SDSERKMDPA EEDTNVYTEK HSDSLFKRTE VL Antigen sequence identity: Mouse 68%; Rat 63%.

Syndecans are heparan sulfate proteoglycans (HSPGs) that are involved in cell adhesion, tissue morphogenesis, differentiation, and act as low affinity co-receptors of soluble growth factors. It is involved in the mitogenic activity and signaling of Granulocyte-Macrophage Colony-stimulating factor in osteoblasts, and has been shown to have a regulatory role in matrix assembly. Syndecan-2 is expressed at epithelial-mesenchymal interfaces, and prechondrogenic and preosteogenic mesenchymal condensations, which are areas of high morphogenetic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan 2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SDC2
Gene Accession No. P34741
Gene Alias 4833414L08Rik; AA960457; CD362; cell surface-associated heparan sulfate proteoglycan 1; fibroglycan; heparan sulfate proteoglycan 1, cell surface-associated; heparan sulfate proteoglycan 1, cell surface-associated (fibroglycan); heparan sulfate proteoglycan core protein; HSPG; Hspg1; Ryudocan/syndecan 2; Sdc2; SYND2; syndecan 2; syndecan proteoglycan 2; syndecan-2
Gene Symbols SDC2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SDC2 (Product #RP-106068)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6383
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.