missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syndecan-2/CD362 Antibody (305515R), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2965RH
This item is not returnable.
View return policy
Description
Syndecan-2/CD362 Monoclonal antibody specifically detects Syndecan-2/CD362 in Human samples. It is validated for Flow Cytometry
Specifications
| Syndecan-2/CD362 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Extracellular Matrix | |
| 6383 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 305515R | |
| Flow Cytometry | |
| CD362 antigen, fibroglycan, heparan sulfate proteoglycan 1, cell surface-associated, Heparan sulfate proteoglycan core protein, HSPG, HSPG1syndecan proteoglycan 2, SYND2cell surface-associated heparan sulfate proteoglycan 1, syndecan 2, syndecan-2 | |
| Mouse myeloma cell line NS0-derived recombinant human Syndecan-2/CD362, Glu19-Gly144, Accession # AAH49836 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Spot an opportunity for improvement?Share a Content Correction