missing translation for 'onlineSavingsMsg'
Learn More

Syndecan-1/CD138 Antibody (320611) [DyLight 405], Novus Biologicals™

Product Code. 30735780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735780 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735780 Supplier Novus Biologicals Supplier No. FAB29661E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syndecan-1/CD138
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 320611
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD138, CD138 antigen, SDC, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Syndecan-1/CD138 isoform 1, Gln18-Glu252 (predicted), Accession # P18828
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers
Primary or Secondary Primary
Gene ID (Entrez) 6382
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.