Learn More
Invitrogen™ Synaptophysin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA11043
Product Code. 11515892
  • £476.00 / 100µg
Description
PA1-1043 detects Synaptophysin in rat, mouse, bovine, and sheep samples. PA1-1043 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a ~33.8 kDa protein representing Synaptophysin in rat, mouse and bovine brain tissue as well as sheep retinas. The PA1-1043 immunogen is a synthetic peptide corresponding to residues G(253) P G G Y G P Q D S Y G P Q G G Y Q P D(272) of rat Synaptophysin. This sequence is conserved in many species including human, mouse and bovine. The PA1-1043 immunizing peptide (Cat. # PEP-279) is available for use in neutralization and control experiments.
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Synaptophysin
Polyclonal
Unconjugated
Syp
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1
Rabbit
Antigen affinity chromatography
RUO
101112034, 20977, 24804, 280937, 6855
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P07825, P08247, P20488, Q62277
Syp
Synthetic peptide corresponding to residues G(253) P G G Y G P Q D S Y G P Q G G Y Q P D(272) of rat Synaptophysin.
100 μg
Primary
Human, Mouse, Rat, Bovine, Sheep