missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Antibody (EP10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£153.00 - £398.00
Specifications
| Antigen | Synaptophysin |
|---|---|
| Clone | EP10 |
| Dilution | Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18218983
|
Novus Biologicals
NBP1-19222-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776433
|
Novus Biologicals
NBP1-19222 |
0.1 mg | |||||||
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Synaptophysin | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat, Human | |
| Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
| SYP | |
| IgG1 | |
| Protein G purified | |
| Synaptophysin (EP10) |
| EP10 | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Membrane Vesicle Markers, Stem Cell Markers | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 6855 | |
| Immunoprecipitate of human brain | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title