missing translation for 'onlineSavingsMsg'
Learn More

Synaptojanin 2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18403217 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403217 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403217 Supplier Novus Biologicals Supplier No. NBP222314AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Synaptojanin 2 Polyclonal antibody specifically detects Synaptojanin 2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synaptojanin 2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.36, inositol phosphate 5'-phosphatase 2, INPP5H, KIAA0348MGC44422, Synaptic inositol-14,5-trisphosphate 5-phosphatase 2, synaptojanin 2, synaptojanin-2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1275 to 1325 of human Synaptojanin 2 using the numbering given in entry NP_003889.1 (GeneID 8871).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 8871
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.