missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synaptojanin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582442
This item is not returnable.
View return policy
Description
Immunogen sequence: ASFGEVILIR FVEDKMWVTF LEGSSALNVL SLNGKELLNR TITIALKSPD WIKNLEEEMS LEKISIALPS STSSTLLGED AEVAADFDME GDVDDYSAEV EELLPQHLQP SSSSGLGTSP SSSPRTSPCQ S Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.
Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.
Specifications
| Synaptojanin 1 | |
| Polyclonal | |
| Unconjugated | |
| SYNJ1 | |
| A930006D20Rik; AA675315; inositol 5'-phosphatase (synaptojanin 1); inositol polyphosphate-5-phosphatase G; INPP5G; KIAA0910; mKIAA0910; PARK20; phosphoinositide 5-phosphatase; synaptic inositol 1,4,5-trisphosphate 5-phosphatase 1; synaptic inositol-1; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1; synaptojanin 1; synaptojanin-1; synaptojanin-1, polyphosphoinositide phosphatase; Synj1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8867 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43426 | |
| SYNJ1 | |
| Recombinant protein corresponding to Human SYNJ1. Recombinant protein control fragment (Product #RP-89906). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction