missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptogyrin 4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-984
This item is not returnable.
View return policy
Description
Synaptogyrin 4 Polyclonal specifically detects Synaptogyrin 4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Synaptogyrin 4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| MGC125805, synaptogyrin 4, synaptogyrin-4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23546 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.01 - 0.1 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| NP_036583. | |
| SYNGR4 | |
| Peptide with sequence HIPKSLQELANS-C corresponding to N-Terminus according to NP_036583.2. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction